missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. 15274744 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15274744 1 g 1g
15264744 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15274744 Supplier Thermo Scientific Acros Supplier No. 442900010
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 368-48-9
Molecular Formula C6H4F3N
Molecular Weight (g/mol) 147.10
MDL Number MFCD02183562
InChI Key ATRQECRSCHYSNP-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyridine, 2-trifluoromethyl-pyridine, trifluoromethylpyridine, 2-1,1,1-trifluoromethyl pyridine, pyridine, trifluoromethyl, alpha,alpha,alpha-trifluoro-2-picoline, 2-cf3-pyridine, trifluoromethyl pyridine
PubChem CID 136199
IUPAC Name 2-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC=CC=N1

Specifications

Boiling Point 138°C to 141°C
Flash Point 42°C
Assay Percent Range 96% min. (GC)
Refractive Index 1.4183
Quantity 1 g
Formula Weight 147.1
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)pyridine
compliance-icons compliance-icons
Product Identifier
  • 2-(Trifluoromethyl)pyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.