missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)quinoline, 97%, Thermo Scientific™

Product Code. 10442878 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10442878 5 g Glass bottle 5g
10349167 1 g Glass bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10442878

Brand: Thermo Scientific Acros 432620050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 347-42-2
Molecular Formula C10H6F3N
Molecular Weight (g/mol) 197.16
MDL Number MFCD01862002
InChI Key YZSRICFIQLVSMQ-UHFFFAOYSA-N
Synonym 2-trifluoromethyl quinoline, 2-trifluoromethyl-quinoline, pubchem23495, trifluoromethyl quinoline, quinoline,2-trifluoromethyl
PubChem CID 2777805
IUPAC Name 2-(trifluoromethyl)quinoline
SMILES FC(F)(F)C1=NC2=CC=CC=C2C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 58°C to 62°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 197.16
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)quinoline
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Trifluoromethyl)quinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.