missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)ethanol, 98+%

Product Code. 11480037
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480037 100 g 100g
11460037 5 g 5g
11470037 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480037 Supplier Thermo Scientific Alfa Aesar Supplier No. B20970.22
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2916-68-9 | C5H14OSi | 118.251 g/mol

2-(Trimethylsilyl)ethanol is used as a protecting reagent for carboxyl, phosphoryl, hydroxyl and amino group in organic synthesis. It is used as a precursor to prepare trimethyl(2-phenoxyethyl)silanes by reacting with aromatic fluoride. It is also used in the synthesis of teoc-protected amines by using the corresponding isocyanates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trimethylsilyl)ethanol is used as a protecting reagent for carboxyl, phosphoryl, hydroxyl and amino group in organic synthesis. It is used as a precursor to prepare trimethyl(2-phenoxyethyl)silanes by reacting with aromatic fluoride. It is also used in the synthesis of teoc-protected amines by using the corresponding isocyanates.

Solubility
Miscible with water.

Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2916-68-9
Molecular Formula C5H14OSi
Molecular Weight (g/mol) 118.251
MDL Number MFCD00002825
InChI Key ZNGINKJHQQQORD-UHFFFAOYSA-N
Synonym 2-trimethylsilyl ethanol, ethanol, 2-trimethylsilyl, 2-hydroxyethyl trimethylsilane, 2-trimethylsilyl ethan-1-ol, silane, 2-hydroxyethyl trimethyl, 2-trimethylsilanyl-ethanol, 2-trimethylsilyl-ethanol, 2-trimethylsilyl ethanal
PubChem CID 18013
IUPAC Name 2-trimethylsilylethanol
SMILES C[Si](C)(C)CCO

Specifications

Density 0.825
Boiling Point 71°C to 72°C (10 mmHg)
Flash Point 50°C (122°F)
Assay Percent Range 98+%
Refractive Index 1.425
Quantity 100 g
UN Number UN1987
Beilstein 1732034
Solubility Information Miscible with water.
Formula Weight 118.25
Percent Purity ≥98%
Chemical Name or Material 2-(Trimethylsilyl)ethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.