missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)ethanol, 98+%

Product Code. 10410681 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10410681 10 mL Glass bottle 10mL
10512364 50 mL Glass bottle 50mL
11995811 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10410681 Supplier Thermo Scientific Acros Supplier No. 210480100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2916-68-9 | C5H14OSi | 118.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2916-68-9
Molecular Formula C5H14OSi
Molecular Weight (g/mol) 118.25
MDL Number MFCD00002825
InChI Key ZNGINKJHQQQORD-UHFFFAOYSA-N
Synonym 2-trimethylsilyl ethanol, ethanol, 2-trimethylsilyl, 2-hydroxyethyl trimethylsilane, 2-trimethylsilyl ethan-1-ol, silane, 2-hydroxyethyl trimethyl, 2-trimethylsilanyl-ethanol, 2-trimethylsilyl-ethanol, 2-trimethylsilyl ethanal
PubChem CID 18013
IUPAC Name 2-trimethylsilylethanol
SMILES C[Si](C)(C)CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.8250g/mL
Boiling Point 71.0°C to 73.0°C (35.0 mmHg)
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)3SiCH2CH2OH
Refractive Index 1.4235 to 1.4255
Quantity 10 mL
Specific Gravity 0.825
Solubility Information Solubility in water: soluble in water
Formula Weight 118.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(Trimethylsilyl)ethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(Trimethylsilyl)ethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.