missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilylethynyl)pyridine, 97%

Product Code. 11357646
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11357646

Brand: Thermo Scientific Alfa Aesar L11251.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86521-05-3 | C10H13NSi | 175.306 g/mol

2-(Trimethylsilylethynyl)pyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trimethylsilylethynyl)pyridine is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86521-05-3
Molecular Formula C10H13NSi
Molecular Weight (g/mol) 175.306
MDL Number MFCD00066349
InChI Key WOFPTESETJKCBH-UHFFFAOYSA-N
Synonym 2-trimethylsilylethynyl pyridine, 2-trimethylsilyl ethynyl pyridine, trimethyl 2-pyridin-2-ylethynyl silane, 2-2-trimethylsilyl ethynyl pyridine, timtec-bb sbb009057, trimethylsilylethynylpyridine, trimethylsilylethynyl pyridin, acmc-1bl66, 2-trimethylsilylethynylpyridine
PubChem CID 522859
IUPAC Name trimethyl(2-pyridin-2-ylethynyl)silane
SMILES C[Si](C)(C)C#CC1=CC=CC=N1

Specifications

Melting Point 40°C to 43°C
Boiling Point 85°C to 86°C (1 mmHg)
Refractive Index 1.533
Quantity 1 g
Beilstein 4176916
Solubility Information Insoluble in water.
Formula Weight 175.31
Percent Purity 97%
Chemical Name or Material 2-(Trimethylsilylethynyl)pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.