missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilylethynyl)pyridine, 97%

Product Code. 11367646
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367646 5 g 5g
11357646 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367646 Supplier Thermo Scientific Alfa Aesar Supplier No. L11251.06
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 86521-05-3 | C10H13NSi | 175.306 g/mol

2-(Trimethylsilylethynyl)pyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trimethylsilylethynyl)pyridine is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86521-05-3
Molecular Formula C10H13NSi
Molecular Weight (g/mol) 175.306
MDL Number MFCD00066349
InChI Key WOFPTESETJKCBH-UHFFFAOYSA-N
Synonym 2-trimethylsilylethynyl pyridine, 2-trimethylsilyl ethynyl pyridine, trimethyl 2-pyridin-2-ylethynyl silane, 2-2-trimethylsilyl ethynyl pyridine, timtec-bb sbb009057, trimethylsilylethynylpyridine, trimethylsilylethynyl pyridin, acmc-1bl66, 2-trimethylsilylethynylpyridine
PubChem CID 522859
IUPAC Name trimethyl(2-pyridin-2-ylethynyl)silane
SMILES C[Si](C)(C)C#CC1=CC=CC=N1

Specifications

Melting Point 40°C to 43°C
Boiling Point 85°C to 86°C (1 mmHg)
Refractive Index 1.533
Quantity 5 g
Beilstein 4176916
Solubility Information Insoluble in water.
Formula Weight 175.31
Percent Purity 97%
Chemical Name or Material 2-(Trimethylsilylethynyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.