missing translation for 'onlineSavingsMsg'
Learn More

2-Undecanone, 98%

Product Code. 10093013 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10093013 1 L Glass bottle 1 L
10225050 5 mL Glass bottle 5mL
10061630 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10093013 Supplier Thermo Scientific Acros Supplier No. 140690010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-12-9 | C11H22O | 170.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-12-9
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.30
MDL Number MFCD00009583
InChI Key KYWIYKKSMDLRDC-UHFFFAOYSA-N
Synonym 2-undecanone, methyl nonyl ketone, 2-hendecanone, ketone, methyl nonyl, undecanone, nonyl methyl ketone, rue ketone, 2-oxoundecane, methyl n-nonyl ketone, methylnonylketone
PubChem CID 8163
ChEBI CHEBI:17700
IUPAC Name undecan-2-one
SMILES CCCCCCCCCC(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 11.0°C to 13.0°C
Color Colorless to Yellow
Density 0.8240g/mL
Boiling Point 231.0°C to 232.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)8COCH3
Refractive Index 1.4280 to 1.4310
Quantity 1 L
Beilstein 01, 713
Merck Index 15, 6177
Specific Gravity 0.824
Solubility Information Solubility in water: insoluble
Formula Weight 170.3
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Undecanone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Undecanone
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to an approved waste d.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.