missing translation for 'onlineSavingsMsg'
Learn More

2-Vinylpyridine (stabilized with 0.1% 4-tert-butylcatechol), TRC

Product Code. 30003826
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003826 10 g 10g
30005311 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003826 Supplier Toronto Research Chemicals Supplier No. TRCV48749510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-69-6
Molecular Formula C7 H7 N
Molecular Weight (g/mol) 105.14
Synonym 2-Ethenylpyridine, Betahistine Dihydrochloride Imp. A (EP), 2-Vinylpyridine
IUPAC Name 2-ethenylpyridine
SMILES C=Cc1ccccn1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H7N/c1-2-7-5-3-4-6-8-7/h2-6H,1H2
Formula Weight 105.0578
Percent Purity >95%
Chemical Name or Material 2-Ethenylpyridine (2-Vinylpyridine)
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.