Learn More
Ohaus™ 2 x Centrifuge Bucket Adapters, 3xMTP, Frontier™ R-S6xMTP/4MI, Swing Out Rotors
Product Code. 15876722 Shop All Ohaus Products
Click to view available options
Capacity:
48 x 1.5/2.0 mL
Unit Size:
Pack of 2
Product Code. 15876722
Brand: Ohaus™ 30314891
Holds a rectangular array of tubes for safe and secure centrifugation in R-S6xMTP/4MI, Swing Out Rotors.
  • Holds a rectangular array/rack of tubes
  • Suitable for use in Frontier™ R-S6xMTP/4MI, Swing Out Rotors
TRUSTED_SUSTAINABILITY
Specifications
Capacity 48 x 1.5/2.0 mL
Max. Tube Diameter 11 mm
For Use With (Equipment) R-S6xMTP/4MI, Swing Out Rotors
Description Adapter, 48x1.5/2.0ml D11mm, 2/pk
No. of Places 48
No. per Pack 2