Learn More
Ohaus™ 2 x Centrifuge Buckets, Frontier™ R-S6x5/4 Swing Out Rotor
Product Code. 15846642 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
5 mL
Unit Size:
Pack of 2
Product Code. Capacity unitSize
15846642 5 mL Pack of 2
1 options
Product Code. 15846642 Supplier Ohaus™ Supplier No. 30130895
Only null left
Add to basket
Add to basket
Carrier buckets for 5 mL centrifuge tubes, suitable for R-S6x5/4 Swing Out Rotors.
  • Swinging bucket holds specifically sized tubes for centrifugation
  • Suitable for use in Frontier™ R-S6x5/4 Swing Out Rotors
TRUSTED_SUSTAINABILITY
Specifications
Description Buckets, For 30130880 x2
Capacity 5 mL
For Use With (Equipment) R-S6x5/4 Rotor
Includes Buckets for swing out rotor (2 pcs.)
No. of Places 1