Learn More
Brannan™ 200 Series Specific Gravity Hydrometers
Product Code. 15949330
Change view
Click to view available options
Unit Size:
Each
Product Code. Unit Size
15949330 Each
1 options
Product Code. 15949330 Supplier Brannan™ Supplier No. 10/500/0
  • ONLINE EXCLUSIVE PRICE
    16.90 €
    15.47 € / Pack of 1
    Save 1.43 €
    8% Off
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Compares the relative density of a given liquid with that of water at 15.6°C (60°F). Brannan™ 200 Series Specific Gravity Hydrometers are manufactured from soda glass, steel shot ballast and synthetic wax resin.
  • 250mm in length
  • Scale subdivision 0.002
  • Accuracy of ± 0.002 specific gravity
  • Includes protective plastic tube
  • Available adjusted for either low or medium surface tension
  • Available for numerous specific gravity ranges
Specifications
Type 200 Series Specific Gravity Hydrometers
Description ISO 650, 916
Material Glass
For Use With (Application) Low surface tension