missing translation for 'onlineSavingsMsg'
Learn More

LMS™ Series 1A Model 201NP Cooled Incubator, 201 L, Pre-coated Aluminum

Product Code. 11813420
Click to view available options
Capacity (Metric):
201 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11813420

Brand: LMS™ 201NP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Temperature Range: -10°C to +50°C

  • Range of cooled incubators with a white stove enamel steel exterior, foam insulation and a white pre-coated aluminum interior
  • Reliable and accurate temperature control
  • Standard and programmable models available
  • Programmable models offer multiple temperature control in multiple time zones
  • Temperature range -10.0°C to +50.0°C, temperature variation with average workload ±0.5°C
  • Self-evaporating condensate drip tray
  • White plastic coated wire shelves
  • Energy efficient and eco-friendly
  • P.I.D. heating and cooling to ensure stable internal temperature
  • Pre-set safety cutout
  • Hermetically sealed refrigeration system
  • Door lock
  • Controls are easy to read and simple to use
  • Full range of optional extras available. including LED lighting, access porthole, inner perspex doors, alarm system, internal socket and chart recorder
  • Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests

Specifications

Certifications/Compliance ISO 9001:2008
Controller Type Programmable
Description Freestanding incubator
Capacity (Metric) 201 L
Chamber Material Pre-coated Aluminum
Convection Type Fan-assisted Circulation
Dimensions (D x W) Shelf 420 mm x 386 mm
Lighting Optional LED
Material Stove Enameled Mild Steel
No. of Shelves 4
Temperature Range (Metric) -10°C to +50°C
Type Refrigerated Incubator
Display Digital
Insulation Foamed polyurethane
Model 201NP
Refrigerant CFC-free
Temperature Stability ±0.1°C
Temperature Uniformity ±0.5°C
Warranty 2 Years
Width (English) 20.5 in.
Width (English) Interior 17.5 in.
Width (Metric) 520 mm
Width (Metric) Interior 445 mm
Height (English) 56.3 in.
Height (English) Interior 37.0 in.
Height (Metric) 1430 mm
Height (Metric) Interior 965 mm
Depth (English) Interior 19.3 in.
Depth (Metric) Interior 490 mm
Weight (Metric) 65 kg
Electrical Requirements 240 V, 50 Hz
Voltage 240 V
Frequency 50/60 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.