missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 20mm Crimp Seal, Red, Magnetic Bimetal Cap, 8mm Center hole, Assembled septum

Product Code. 16398377 Shop All Fisherbrand Products
Click to view available options
Color:
Beige, White, Red
Blue Transparent, Transparent, Red
Gray, Red
Material (Liner):
PTFE/Butyl
Silicone/PTFE
Thickness (Metric):
3 mm
3.2 mm
Unit Size:
Pack of 100
Pack of 1000

Product Code. 16398377

Only null left
Add to basket
Add to basket

Pre-assembled caps and septa are convenient and minimize contamination from handling.

  • 20mm
  • Closure
  • Magnetic
  • Bi-Metal Cap
  • Red
  • 8mm center hole
  • Combination Seal
TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) PTFE/Butyl
Color Gray, Red
Diameter (Metric) Outer 20 mm
Material Bi-metal
For Use With (Application) Headspace
Quantity 100
Thickness (Metric) 3 mm
Type Crimp Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.