missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 20mm Crimp Seal, Silver, Aluminum Cap, 10mm Center Hole, Assembled Septum

Product Code. 11590505 Shop All Fisherbrand Products
Click to view available options
Color:
Beige, White
Blue, White
Dark Gray
Red-Orange, Transparent
Silver, Blue Transparent, White
Silver, Blue, Gray
Silver, Gray, Red
Silver, White
Material (Liner):
Butyl
Natural Rubber/TEF
PTFE/Butyl
Silicone/Aluminum
Silicone/PTFE
Thickness (Metric):
1.3 mm
1.5 mm
3 mm
3.2 mm
Unit Size:
Pack of 1000

Product Code. 11590505

Only null left
Add to basket
Edge
Add to basket

Pre-assembled caps and septa are convenient and minimize contamination from handling.

  • 20mm
  • Combination Seal
  • aluminum
  • Cap
  • li>Center hole
TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) Silicone/PTFE
Color Beige, White
Diameter (Metric) Outer 20 mm
Material Aluminum
For Use With (Application) Headspace
Quantity 1000
Thickness (Metric) 3.2 mm
Product Type Crimp Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.