missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 20mm Crimp Seal, Silver, Center Tear-Off Cap, assembled septum

Product Code. 11865030 Shop All Fisherbrand Products
Change view
Click to view available options
Material (Liner):
Butyl
PTFE/Butyl
Silicone/PTFE
Color:
Blue Transparent, White
Blue, Gray
Dark Gray
Gray
Thickness (Metric):
3 mm
Unit Size:
Pack of 1000
Product Code. Material (Liner) Color Thickness (Metric) unitSize
11865030 Butyl Dark Gray 3 mm Pack of 1000
11825030 PTFE/Butyl Blue, Gray 3 mm Pack of 1000
11855030 PTFE/Butyl Gray 3 mm Pack of 1000
10666364 Silicone/PTFE Blue Transparent, White 3 mm Pack of 1000
4 options

Product Code. 11865030

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Pre-assembled caps and septa are convenient and minimize contamination from handling.

  • 20mm
  • aluminum Cap
  • Combination Seal
TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) Butyl
Color Dark Gray
Diameter (Metric) Outer 20 mm
Material Aluminum
For Use With (Application) Headspace
Quantity 100
Thickness (Metric) 3 mm
Product Type Crimp Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.