Learn More
Justrite™ 22 Gallon Sure-Grip™ EX Slimline Corrosives/Acid Steel Safety Cabinet With Manual-Closing Doors
Description
Justrite blue steel Slimline safety cabinets for corrosives have the same quality safety features as the cabinets for flammables. All models come with double-wall construction, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self-latching doors and leveling feet. Manual-close door(s) opens to a full 180 degrees and self-latches when pushed closed.
Specifications
Specifications
| Description | Cold-rolled Steel (18 ga) |
| Dimensions (D x W x H) | 18 x 23.25 x 65 in. |
| Door Style | Manual-close |
| Load Bearing Capacity | 22 gal. |
| Color | Blue |
| Material | Steel (CR) |