missing translation for 'onlineSavingsMsg'
Learn More

24-Hydroxybudesonide (Mixture of Diastereomers), TRC

Product Code. 30133744
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133744 25 mg 25mg
30008381 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133744 Supplier Toronto Research Chemicals Supplier No. TRCH83037025MG
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C25H34O7
Molecular Weight (g/mol) 446.53
Synonym (11β,16α)-11,21-Dihydroxy-16,17-[(3-hydroxybutylidene)bis(oxy)]pregna-1,4-diene-3,20-dione
SMILES O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2([H])[C@@H](O)C[C@@]4(C)[C@@]3([H])C[C@@H]5C4(C(CO)=O)OC(CC(O)C)O5)=C1

Specifications

Quantity 25 mg
Chemical Name or Material 24-Hydroxybudesonide (Mixture of Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.