missing translation for 'onlineSavingsMsg'
Learn More
Learn More
24(RS)-Hydroxycholesterol-25,26,26,26,27,27,27-d7, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 24-Hydroxycholesterol-25,26,26,26,27,27,27-d7 |
| CAS | 144154-78-9 |
| Molecular Formula | C27 2H7 H39 O2 |
| Quantity | 0.005 g |
| Synonym | Cholest-5-ene-25,26,26,26,27,27,27-d7-3,24-diol, (3β)-, (3β)-Cholest-5-ene-25,26,26,26,27,27,27-d7-3,24-diol, 24(R/S)-(25,26,26,26,27,27,27-2H7)Hydroxycholesterol, 24(RS)-Hydroxycholesterol-d7, 24-Hydroxycholesterol-25,26,26,26,27,27,27-D7 |
| InChI Formula | InChI=1 S/C27H46O2/c1-17(2)25(29)11-6-18(3)22-9-10-23-21-8-7-19-16-20(28)12-14-26(19,4)24(21)13-15-27(22,23)5/h7,17-18,20-25,28-29 H,6,8-16H2,1-5H3/t18-,20+,21+,22-,23+,24+,25?,26+,27-/m1/s1/i1D3,2D3,17 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C(O)CC[C@@H](C)[C@H]1 CC[C@H]2 [C@@H]3 CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C)C([2 H])([2 H])[2 H] |
| IUPAC Name | (3 S,8 S,9 S,10 R,13 R,14 S,17 R)-10,13-dimethyl-17-[(2 R)-6,7,7,7-tetradeuterio-5-hydroxy-6-(trideuteriomethyl)heptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-3-ol |
| Molecular Weight (g/mol) | 409.7 |
| Formula Weight | 409.3937 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction