missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 24mm PP Screw Seal, Bonded, White, 24-400 Thread, Assembled Septum

Product Code. 10729454 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Product Type:
Center Hole Screw Cap
Solid Top Screw Cap
Unit Size:
Pack of 100
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10729454 Center Hole Screw Cap Pack of 100
10132322 Solid Top Screw Cap Pack of 100
11778266 Solid Top Screw Cap Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10729454
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Pre-assembled caps and septa are convenient and minimize contamination from handling.

  • Ready to use combination seals
  • No time consuming and tricky assembly
  • No contamination of the liner with sweat/fat normally caused by manual assembly
  • White 24-400 caps
TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) Silicone/PTFE
Color Beige, Natural, White
Diameter (Metric) Outer 24 mm
Material Polypropylene
For Use With (Application) Storage
Quantity 100
Thickness (Metric) 3.2 mm
Closure Size 24 mm
For Use With (Equipment) 24-400 Screw Thread Vials
Product Type Center Hole Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.