missing translation for 'onlineSavingsMsg'
Learn More
Learn More
25-Hydroxy Vitamin D2, TRC
Specifications
Specifications
| Chemical Name or Material | 25-Hydroxyvitamin D2 |
| CAS | 21343-40-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C28 H44 O2 |
| Quantity | 0.25 mg |
| Synonym | Cyclohexanol, 4-methylene-3-[(2E)-2-[(1R,3aS,7aR)-octahydro-1-[(1R,2E,4S)-5-hydroxy-1,4,5-trimethyl-2-hexenyl]-7a-methyl-4H-inden-4-ylidene]ethylidene]-, (1S,3Z)-, 9,10-Secoergosta-5,7,10(19),22-tetraene-3,25-diol, (3β,5Z,7E,22E)- (9CI), 9,10-Secoergosta-5,7,10(19),22-tetraene-3β,25-diol (8CI), (1S,3Z)-4-Methylene-3-[(2E)-2-[(1R,3aS,7aR)-octahydro-1-[(1R,2E,4S)-5-hydroxy-1,4,5-trimethyl-2-hexenyl]-7a-methyl-4H-inden-4-ylidene]ethylidene]cyclohexanol, 25-Hydroxycalciferol, 25-Hydroxyergocalciferol, 25-Hydroxyvitamin D, 25-Hydroxyvitamin D2, Ercalcidiol, Vitamin D2-25-hydroxy |
| InChI Formula | InChI=1S/C28H44O2/c1-19-10-14-24(29)18-23(19)13-12-22-8-7-17-28(6)25(15-16-26(22)28)20(2)9-11-21(3)27(4,5)30/h9,11-13,20-21,24-26,29-30H,1,7-8,10,14-18H2,2-6H3/b11-9+,22-12+,23-13-/t20-,21+,24+,25-,26+,28-/m1/s1 |
| SMILES | C[C@H](\C=C\[C@H](C)C(C)(C)O)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)CCC3=C)\CCC[C@]12C |
| IUPAC Name | (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(E,2R,5S)-6-hydroxy-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol |
| Molecular Weight (g/mol) | 412.65 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?