missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Lenalidomide 4'-alkylC3-azide
GREENER_CHOICE

Product Code. 18768036
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18768036 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18768036 Supplier Tocris Bioscience™ Supplier No. 7516/25
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Cereblon ligand with alkyl linker and terminal azide for onward chemistry

Lenalidomide 4'-alkylC3-azide is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC3 linker with terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[4-[(3-Azidopropyl)amino]-1,3-dihydro-1-oxo-2 H-isoindol-2-yl]-2,6-piperidinedione
CAS 2399455-71-9
Quantity 25 mg
Target Degrader Building Blocks
Molecular Formula C16H18N6O3
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.