missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RGFP 966
GREENER_CHOICE

Product Code. 18789826
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18789826

Brand: Tocris Bioscience™ 6728/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective HDAC3 inhibitor (IC50 = 80 nM)

RGFP 966 is a potent and selective HDAC3 inhibitor (IC50 = 80 nM); exhibits no effect on other HDAC (up to 15 μM). RGFP 966 reduces the frequency of striatal CAG repeat expansions in mouse models of Huntington's disease and reduces mHTT accumulation. RGFP 966 enhances long-term object-location memory in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2E)-N-(2-Amino-4-fluorophenyl)-3-[1-(3-phenyl-2-propen-1-yl)-1H-pyrazol-4-yl]-2-propenamide
CAS 1357389-11-7
Quantity 25 mg
Molecular Formula C21H19FN4O
Formula Weight 362.4
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.