missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Thalidomide 4'-ether-PEG2-azide

Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Thalidomide 4'-ether-PEG2-azide is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG2 linker with azide terminal ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 4-(2-(2-(2-Azidoethoxy)ethoxy)ethoxy)-2-(2,6-dioxopiperidin-3-yl)-1 H-isoindole-1,3(2 H)-dione |
| Quantity | 25 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C19H21N5O7 |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu