Learn More
Thermo Scientific™ 2500μL Luer-Tip, Gas-Tight Syringe
Product Code. 15284593 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
2.5 mL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
15284593 2.5 mL Each
1 options
Product Code. 15284593 Supplier Thermo Scientific™ Supplier No. 365L7818
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Thermo Scientific Luer Syringes offer easy, accurate and reproducible injections for a variety of volumes.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Gas Tight GC Autosampler Syringe, Luer Tip
Volume (Metric) 2.5 mL
For Use With (Equipment) Gas Chromatography Systems