missing translation for 'onlineSavingsMsg'
Learn More

Benzyl-d7 Chloride, TRC

Product Code. 30334866
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30334866

Brand: Toronto Research Chemicals TRCB536598250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl-d7 Chloride
CAS 59502-05-5
Molecular Formula C7 D7 Cl
Quantity 250 mg
Synonym Benzene-1,2,3,4,5-d5, 6-(chloromethyl-d2)- (ACI), 6-(Chloromethyl-d2)benzene-1,2,3,4,5-d5 (ACI), Benzene-d5, (chloromethyl-d2)- (9CI), 1-[Chloro(dideuterio)methyl]-2,3,4,5,6-pentadeuteriobenzene, Benzyl chloride-d7, Benzyl-d7 chloride, d7-Benzyl chloride, Perdeuteriobenzyl chloride, Benzyl Chloride-D7
InChI Formula InChI=1S/C7H7Cl/c8-6-7-4-2-1-3-5-7/h1-5H,6H2/i1D,2D,3D,4D,5D,6D2
SMILES [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])Cl
IUPAC Name 1-[chloro(dideuterio)methyl]-2,3,4,5,6-pentadeuterio-benzene
Molecular Weight (g/mol) 133.63
Formula Weight 133.0676
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.