missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bicalutamide Sulfide, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | (2RS)-N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfanyl]-2-hydroxy-2-methylpropanamide |
| CAS | 90356-78-8 |
| Molecular Formula | C18 H14 F4 N2 O2 S |
| Quantity | 250 mg |
| Synonym | N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)thio]-2-hydroxy-2-methylpropanamide, Propanamide, N-[4-cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)thio]-2-hydroxy-2-methyl-, (±)-, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)thio]-2-hydroxy-2-methylpropanamide, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenylsulfanyl)-2-hydroxy-2-methylpropionamide |
| InChI Formula | InChI=1S/C18H14F4N2O2S/c1-17(26,10-27-14-6-3-12(19)4-7-14)16(25)24-13-5-2-11(9-23)15(8-13)18(20,21)22/h2-8,26H,10H2,1H3,(H,24,25) |
| SMILES | CC(O)(CSc1ccc(F)cc1)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F |
| IUPAC Name | N-[4-cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorophenyl)sulfanyl-2-hydroxy-2-methylpropanamide |
| Molecular Weight (g/mol) | 398.37 |
| Formula Weight | 398.0712 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction