missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) Phthalate, TRC

Product Code. 30340775
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 30340775

Brand: Toronto Research Chemicals TRCB433850250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-2-ethylhexyl ester
CAS 117-81-7
Purity Grade Notes HPLC
Molecular Formula C24 H38 O4
Quantity 250 mg
Synonym 1,2-Benzenedicarboxylic acid, bis(2-ethylhexyl) ester (9CI), Phthalic acid, bis(2-ethylhexyl) ester (6CI,8CI), 2-Ethylhexyl phthalate, 3315AF2, Bis(2-ethylhexyl) 1,2-benzenedicarboxylate, Bis(2-ethylhexyl) o-phthalate, Bis(2-ethylhexyl) phthalate, Bisoflex 81, Bisoflex DOP, Codan Set L 86P, Compound 889, Corflex 400, DEHP, DOF, DOF (Russian plasticizer), DOP, DOPT, Di(2-ethylhexyl) phthalate, Di(isooctyl) phthalate, Diacizer DOP, Dioctyl phthalate, Diplast O, ESBO-D 82, Ergoplast FDO, Ergoplast FDO-S, Etalon, Etalon (plasticizer), Ethylhexyl phthalate, Eviplast 80, Eviplast 81, Fleximel, Flexol DOD, Flexol DOP, Garbeflex DOP-D 40, Good-rite GP 264, Hatco DOP, JSSD-DOP, Jayflex DOP, Kodaflex DEHP, Kodaflex DOP, Monocizer DOP, NSC 17069, Octoil, Octyl phthalate, Palatinol AH, Palatinol AH-L, Palatinol DOP, Phthalic acid di(2-ethylhexyl) ester, Phthalic acid dioctyl ester, Pittsburgh PX 138, Plasthall DOP, R 8000, Reomol D 79P, Sansocizer DOP, Sansocizer R 8000, Scandinol SC 1000, Sconamoll DOP, Sicol 150, Staflex DOP, Truflex DOP, Vestinol AH, Vinycizer 80, Vinycizer 80K, Witcizer 312, ZS, ZS (plasticizer)
InChI Formula InChI=1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3
SMILES CCCCC(CC)COC(=O)c1ccccc1C(=O)OCC(CC)CCCC
IUPAC Name bis(2-ethylhexyl) benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 390.56
Formula Weight 390.277
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.