missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-trans-1,2-Bis(2-mercaptoacetamido)cyclohexane, TRC

Product Code. 30340553
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30340553

Brand: Toronto Research Chemicals TRCB481450250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (+/-)-trans-1,2-Bis(2-mercaptoacetamido)cyclohexane
CAS 257641-01-3
Molecular Formula C10 H18 N2 O2 S2
Quantity 250 mg
InChI Formula InChI=1S/C10H18N2O2S2/c13-9(5-15)11-7-3-1-2-4-8(7)12-10(14)6-16/h7-8,15-16H,1-6H2,(H,11,13)(H,12,14)/t7-,8-/m1/s1
SMILES SCC(=O)N[C@@H]1CCCC[C@H]1NC(=O)CS
IUPAC Name 2-sulfanyl-N-[(1R,2R)-2-[(2-sulfanylacetyl)amino]cyclohexyl]acetamide
Molecular Weight (g/mol) 262.39
Formula Weight 262.081
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.