missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Amino-1-oxoisoindolin-2-yl)-4-carbamoyl Butyric Acid, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | 4-(4-Amino-1-oxoisoindolin-2-yl)-4-carbamoyl Butyric Acid |
| CAS | 2197414-57-4 |
| Molecular Formula | C13 H15 N3 O4 |
| Quantity | 25 mg |
| Synonym | 2-(4-Amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)pentanedioic acid, 5-Amino-4-(4-amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)-5-oxopentanoic Acid |
| InChI Formula | InChI=1S/C13H15N3O4/c14-9-3-1-2-7-8(9)6-16(13(7)20)10(12(15)19)4-5-11(17)18/h1-3,10H,4-6,14H2,(H2,15,19)(H,17,18) |
| SMILES | NC(=O)C(CCC(=O)O)N1Cc2c(N)cccc2C1=O |
| IUPAC Name | 5-amino-4-(4-amino-1-oxo-isoindolin-2-yl)-5-oxo-pentanoic acid |
| Molecular Weight (g/mol) | 277.28 |
| Formula Weight | 277.1063 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction