missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopenthiazide, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Cyclopenthiazide |
| CAS | 742-20-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13 H18 Cl N3 O4 S2 |
| Quantity | 25 mg |
| Synonym | 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 6-chloro-3-(cyclopentylmethyl)-3,4-dihydro-, 1,1-dioxide, 6-Chloro-3-(cyclopentylmethyl)-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, 6-Chloro-3-(cyclopentylmethyl)-3,4-dihydro-7-sulfamyl-2H-1,2,4-benzothiadiazine 1,1-dioxide, 6-Chloro-3-cyclopentylmethyl-7-sulfamoyl-3,4-dihydro-1,2,4-benzothiadiazine 1,1-dioxide, Benesal, Ciba 8341-Su, Cyclomethiazide, Cyclometiazid, Cyclopenthiazide, Cyclopentiazid, NSC 107679, Navidreks, Navidrex, Navidrix, Salimed, Salimid, Salurilo-C, Su 8341, Tsiklometiazid, Ultra-Minzil |
| InChI Formula | InChI=1S/C13H18ClN3O4S2/c14-9-6-10-12(7-11(9)22(15,18)19)23(20,21)17-13(16-10)5-8-3-1-2-4-8/h6-8,13,16-17H,1-5H2,(H2,15,18,19) |
| SMILES | NS(=O)(=O)c1cc2c(NC(CC3CCCC3)NS2(=O)=O)cc1Cl |
| IUPAC Name | 6-chloro-3-(cyclopentylmethyl)-1,1-dioxo-3,4-dihydro-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide |
| Molecular Weight (g/mol) | 379.88 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction