missing translation for 'onlineSavingsMsg'
Learn More

Ipamorelin Acetate, TRC

Product Code. 30005845
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005845 25 mg 25mg
30137490 100 mg 100mg
30008297 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005845 Supplier Toronto Research Chemicals Supplier No. TRCI73941525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Synonym 2-Methylalanyl-L-histidyl-3-(2-naphthalenyl)-D-alanyl-D-phenylalanyl-L-lysinamide acetate∣696: PN: WO2004005342 PAGE: 46 claimed protein∣NNC 26-0161∣Sequence Length: 5 modified

Specifications

Format Neat
Molecular Weight (g/mol) 711.856005
Product Type Metabolite
Quantity 25 mg
Shipping Condition Room Temperature
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.