missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-Anabasine (>98%), TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | 3-[(2S)-Piperidin-2-yl]pyridine (Anabasine) |
| CAS | 494-52-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 H14 N2 |
| Quantity | 25 mg |
| Synonym | 3-[(2S)-Piperidin-2-yl]pyridine, Nicotine Imp. G (EP), Anabasine, Pyridine, 3-(2S)-2-piperidinyl-, Anabasine (8CI), Pyridine, 3-(2-piperidinyl)-, (S)-, 3-(2S)-2-Piperidinylpyridine, (-)-2-(3'-Pyridyl)piperidine, (-)-Anabasine, (2S)-2-(Pyridin-3-yl)piperidine, (S)-(-)-Anabasine, (S)-Anabasine, Anabasin, NSC 87504, Neonicotine, Neonikotin |
| InChI Formula | InChI=1S/C10H14N2/c1-2-7-12-10(5-1)9-4-3-6-11-8-9/h3-4,6,8,10,12H,1-2,5,7H2/t10-/m0/s1 |
| SMILES | C1CC[C@H](NC1)c2cccnc2 |
| IUPAC Name | 3-[(2S)-piperidin-2-yl]pyridine |
| Molecular Weight (g/mol) | 162.23 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction