Learn More
3M™ 2840 Series Polycarbonate Lens Safety Glasses
Product Code. 12960305
Click to view available options
Lens Tint:
Amber
Clear
Mirror
Unit Size:
Each
Pack of 20
Product Code. 12960305
Brand: 3M™ 7000032459
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certifications/Compliance EN 166:2001
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety glasses; Safety Spectacles
For Use With (Application) Construction,‎ pharmaceuticals
Frame Color Silver
No. per Pack 20/Pk.
Temple Style Adjustable length
Lens Material Polycarbonate