Learn More
3M™ 2890 Series Polycarbonate Lens Safety Goggles
Product Code. 12672605 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Anti-fog, Scratch-resistant
Scratch-resistant
Ventilation Details:
Indirect ventilation
Sealed
Unit Size:
Each
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Coating Ventilation Details Unit Size
12672605 Anti-fog Sealed Each
12642605 Anti-fog, Scratch-resistant Sealed Each
12652605 Anti-fog Indirect ventilation Each
10361910 Scratch-resistant Indirect ventilation Each
13458997 Scratch-resistant Indirect ventilation Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12672605 Supplier 3M™ Supplier No. 7000032483
  • ONLINE EXCLUSIVE PRICE
    28.15 €
    15.95 € / Pack of 1
    Save 12.20 €
    43% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Certifications/Compliance EN 166:2001
Lens Coating Anti-fog
Lens Tint Clear
Product Type Safety Goggles
For Use With (Application) Construction,‎ laboratories
No. per Pack 1 Ea.
Ventilation Details Sealed
UV Protection Yes