missing translation for 'onlineSavingsMsg'
Learn More

(2E)-3-[[(4-Methylphenyl)sulfonyl]oxy]-2-butenoic Acid Methyl Ester, TRC

Codice prodotto. 30475531
Cambia vista
Click to view available options
Quantity:
500 mg
Dimensione della confezione:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
30475531 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Product Code. 30475531 Supplier Toronto Research Chemicals Supplier No. TRCM327450500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2E)-3-(((4-Methylphenyl)sulfonyl)oxy)-2-butenoic Acid Methyl Ester
CAS 1029612-16-5
Molecular Formula C12H14O5S
Quantity 500 mg
InChI Formula InChI=1S/C12H14O5S/c1-9-4-6-11(7-5-9)18(14,15)17-10(2)8-12(13)16-3/h4-8H,1-3H3/b10-8+
SMILES CC1=CC=C(C=C1)S(=O)(=O)O/C(=C/C(=O)OC)/C
Molecular Weight (g/mol) 270.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.