missing translation for 'onlineSavingsMsg'
Learn More

2H-Perfluoro-5,8,11,14-tetramethyl-3,6,9,12,15-pentaoxaoctadecane, Thermo Scientific™

Product Code. 11373846
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373846 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11373846 Supplier Thermo Scientific Alfa Aesar Supplier No. L16952.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 37486-69-4
Melting Point -95°C
Density 1.765
Boiling Point 221°C to 225°C
Flash Point None
Molecular Formula C17HF35O5
MDL Number MFCD00054716
Quantity 5 g
Synonym 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,16,16,17,17,18,18,18-tricosafluoro-5,8,11,14-tetrakis trifluoromethyl-3,6,9,12,15-pentaoxaoctadecane, 2h-perfluoro-5,8,11,14-tetramethyl-3,6,9,12,15-pentaoxaoctadecane, 3,6,9,12,15-pentaoxaoctadecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,16,16,17,17,18,18,18-tricosafluoro-5,8,11,14-tetrakis trifluoromethyl, 1,1,1,2,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy propoxy-3-1,2,2,2-tetrafluoroethoxy propane, unii-sef36f005e, freon e-5, 2h-tricosafluoro-5,8,11,14-tetrakis trifluoromethyl-3,6,9,12,15-pentaoxaoctadecane, 1,1,1,2,3,3-hexakis fluoranyl-2-1,1,2,3,3,3-hexakis fluoranyl-2-1,1,2,3,3,3-hexakis fluoranyl-2-1,1,2,3,3,3-hexakis fluoranyl-2-1,1,2,2,3,3,3-heptakis fluoranyl propoxy propoxy propoxy propoxy-3-1,2,2,2-tetrakis fluoranyl ethoxy propane
InChI Key MEXQRXXROOSHGK-UHFFFAOYNA-N
SMILES FC(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F
IUPAC Name 1,1,1,2,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy]propoxy]-3-(1,2,2,2-tetrafluoroethoxy)propane
Molecular Weight (g/mol) 950.13
PubChem CID 123459
Formula Weight 950.14
Chemical Name or Material 2H-Perfluoro-5,8,11,14-tetramethyl-3,6,9,12,15-pentaoxaoctadecane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.