missing translation for 'onlineSavingsMsg'
Learn More

(2R)-2-[(S)-hydroxyphenylmethyl]-4-morpholinecarboxylic Acid 1,1-Dimethylethyl Ester, TRC

Product Code. 30406334
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406334 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406334 Supplier Toronto Research Chemicals Supplier No. TRCH95024025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-2-[(S)-hydroxyphenylmethyl]-4-morpholinecarboxylic Acid 1,1-Dimethylethyl Ester
CAS 1030837-58-1
Molecular Formula C16H23NO4
Quantity 25 mg
Synonym (R)-t-Butyl 2-((S)-Hydroxy(phenyl)methyl)morpholine-4-carboxylate
InChI Formula InChI=1S/C16H23NO4/c1-16(2,3)21-15(19)17-9-10-20-13(11-17)14(18)12-7-5-4-6-8-12/h4-8,13-14,18H,9-11H2,1-3H3/t13-,14+/m1/s1
SMILES CC(C)(C)OC(=O)N1CCO[C@H](C1)[C@@H](O)c2ccccc2
IUPAC Name tert-butyl (2R)-2-[(S)-hydroxy(phenyl)methyl]morpholine-4-carboxylate
Molecular Weight (g/mol) 293.36
Formula Weight 293.1627
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.