missing translation for 'onlineSavingsMsg'
Learn More

(2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triol, 97%, BLD Pharm™

Product Code. 30797118
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30797118 1 g 1g
30792146 250 mg 250mg
30796630 25 g 25g
30788814 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30797118 Supplier BLD Pharm Supplier No. BD1475291g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 16758-34-2
Molecular Formula C12H16O5S
MDL Number MFCD00063272
Quantity 1 g
SMILES O[C@H]([C@H]([C@H]([C@@H](CO)O1)O)O)[C@@H]1SC2=CC=CC=C2
Molecular Weight (g/mol) 272.32
Percent Purity 97%
Chemical Name or Material (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.