missing translation for 'onlineSavingsMsg'
Learn More

(2R,3R)-(+)-Bis(diphenylphosphino)butane, 98%

Product Code. 15473687
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473687 250 mg 250mg
11377299 50 mg 50mg
11387299 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15473687 Supplier Thermo Scientific Alfa Aesar Supplier No. H27448.MD
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 74839-84-2 | C28H28P2 | 426.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74839-84-2
Molecular Formula C28H28P2
Molecular Weight (g/mol) 426.48
MDL Number MFCD00075403
InChI Key FWXAUDSWDBGCMN-ZBXWWWQKNA-N
Synonym r,r-chiraphos, chiraphos, r,r, unii-1t086b9q1j, 2r,3r-+-bis diphenylphosphino butane, 2r,3r-butane-2,3-diylbis diphenylphosphine, 2r,3r-+-2,3-bis diphenylphosphino butane, cira, phosphine, 1r,2r-1,2-dimethyl-1,2-ethanediyl bis diphenyl, 2r,3r-bis diphenylphosphino butane, r,?r ?-?chiraphos
PubChem CID 2724948
IUPAC Name [(2R,3R)-3-diphenylphosphanylbutan-2-yl]-diphenylphosphane
SMILES C[C@H]([C@@H](C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 104°C to 109°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 426.48
Percent Purity 98%
Chemical Name or Material (2R,3R)-(+)-Bis(diphenylphosphino)butane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.