missing translation for 'onlineSavingsMsg'
Learn More

(2R,3S)-1-Carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt, 85%, Tech., Thermo Scientific™

Product Code. 10334412 Shop All Thermo Scientific Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10334412

Brand: Thermo Scientific 371002500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205639-93-6
MDL Number MFCD00142975
InChI Key BBLPZPQHEWQLGM-VZXYPILPSA-M
Synonym potassium 5s,6r-5,6-dihydroxy-4-pentylcyclohexa-1,3-dienecarboxylate, 2r,3s-1-carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt
PubChem CID 23666349
IUPAC Name potassium;(5S,6R)-5,6-dihydroxy-4-pentylcyclohexa-1,3-diene-1-carboxylate
SMILES CCCCCC1=CC=C(C(C1O)O)C(=O)[O-].[K+]

Specifications

CAS Min % 80.0
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 80% min. (HPLC)
Packaging Glass bottle
Molecular Formula C12H17KO4
Quantity 250 mg
Molecular Weight (g/mol) 264.35
Formula Weight 264.35
Percent Purity 85%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material (2R,3S)-1-Carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene potassium salt, tech., 85%
Show More Show Less
compliance-icons
Product Identifier
  • (2R,3S)-1-Carboxy-4-pentyl-2,3-dihydroxycyclohexa-4,6-diene, potassium salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.