missing translation for 'onlineSavingsMsg'
Learn More

(2R,3S)-N-Benzoyl-3-phenyl Isoserine Methyl Ester, TRC

Product Code. 30347780
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347780 1 g 1g
30329594 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30347780 Supplier Toronto Research Chemicals Supplier No. TRCB2085601G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl (2R,3S)-3-Benzamido-2-hydroxy-3-phenylpropanoate
CAS 32981-85-4
Molecular Formula C17 H17 N O4
Quantity 1 g
Synonym Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, methyl ester, (αR,βS)-, Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, methyl ester, [R-(R*,S*)]-, Isoserine, N-benzoyl-3-phenyl-, methyl ester, (2R,3S)- (8CI), (2R,3S)-Methyl 3-benzoylamino-2-hydroxy-3-phenylpropanoate, (2R,3S)-N-Benzoyl-3-phenylisoserine methyl ester, LKT-T 0104, Methyl (-)-(2R,3S)-3-benzoylamino-2-hydroxy-3-phenylpropionate, Methyl (2R,3S)-N-benzoylphenylisoserinate
InChI Formula InChI=1S/C17H17NO4/c1-22-17(21)15(19)14(12-8-4-2-5-9-12)18-16(20)13-10-6-3-7-11-13/h2-11,14-15,19H,1H3,(H,18,20)/t14-,15+/m0/s1
SMILES COC(=O)[C@H](O)[C@@H](NC(=O)c1ccccc1)c2ccccc2
IUPAC Name methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropanoate
Molecular Weight (g/mol) 299.32
Formula Weight 299.1158
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.