missing translation for 'onlineSavingsMsg'
Learn More

(+)-(2R,8aS)-(Camphorylsulfonyl)oxaziridine, 98+%

Product Code. 10561711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10561711 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10561711 Supplier Thermo Scientific Acros Supplier No. 294580010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 104322-63-6 | C10H15NO3S | 229.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 104322-63-6
CAS Min % 98.0
CAS Max % 100.0
Melting Point 170.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C10H15NO3S
MDL Number MFCD00075319
Quantity 1 g
Fieser 16,61; 17,66
Specific Rotation Condition 46.60 (25.00°C c=2,CHCl3)
Specific Rotation 46.6
Synonym 1r---10-camphorsulfonyl oxaziridine
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform
InChI Key GBBJBUGPGFNISJ-SKVWKECBSA-N
SMILES CC1(C2CCC13CS(=O)(=O)N4C3(C2)O4)C
Molecular Weight (g/mol) 229.3
PubChem CID 86278372
Formula Weight 229.3
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (+)-(2R, 8aS)-(Camphorylsulfonyl)oxaziridine, 99+%, (99+% e.e.)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.