missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S)-2-Amino-3-phenylpropionyl Amide, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | (2S)-2-Amino-3-phenylpropionyl Amide |
| CAS | 5241-58-7 |
| Molecular Formula | C9H12N2O |
| Quantity | 10 g |
| Synonym | (alphaS)-alpha-Amino-benzenepropanamide, (S)-alpha-Amino-hydrocinnamamide, L-Phenylalaninamide, L-Phenylalanine Amide |
| InChI Formula | InChI=1S/C9H12N2O/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H2,11,12)/t8-/m0/s1 |
| SMILES | N[C@@H](Cc1ccccc1)C(=O)N |
| IUPAC Name | (2S)-2-amino-3-phenylpropanamide |
| Molecular Weight (g/mol) | 164.2 |
| Formula Weight | 164.095 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction