missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-2-Amino-3-methoxybutanoic acid, 98%

Product Code. 11935421 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11935421 1 g Glass bottle 1g
10581691 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11935421 Supplier Thermo Scientific Acros Supplier No. 290030010
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 104195-80-4 | C5H11NO3 | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 104195-80-4
Molecular Formula C5H11NO3
Molecular Weight (g/mol) 133.15
InChI Key FYCWLJLGIAUCCL-IMJSIDKUSA-N
Synonym 2s,3s-2-amino-3-methoxybutanoic acid, 2s,3s-2-amino-3-methoxy-butanoic acid, allo-o-methyl-l-thr, l-allo-o-methylthreonine, 2s,3s-2-amino-3-methoxybutanoic acid
PubChem CID 151207
IUPAC Name (2S,3S)-2-amino-3-methoxybutanoic acid
SMILES CC(C(C(=O)O)N)OC

Spécification

Melting Point 220.0°C to 224.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3CH(OCH3)CH(NH2)COOH
Quantity 1 g
Specific Rotation Condition + 27.50 (25.00°C c=0.6,6N HCl)
Specific Rotation + 27.50
Solubility Information Solubility in water: soluble in water.
Formula Weight 133.15
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material (2S, 3S)-2-Amino-3-methoxybutanoic acid
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.