missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-(-)-Bis(diphenylphosphino)butane, 98%

Product Code. 15413727
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15413727 250 mg 250mg
11476490 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15413727

Brand: Thermo Scientific Alfa Aesar H27586.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 64896-28-2 | C28H28P2 | 426.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64896-28-2
Molecular Formula C28H28P2
Molecular Weight (g/mol) 426.48
MDL Number MFCD00009836
InChI Key FWXAUDSWDBGCMN-ZBXWWWQKNA-N
Synonym s,s-chiraphos, 2s,3s---bis diphenylphosphino butane, chiraphos, chiraphos, s,s, unii-6qr78gzl9b, 6qr78gzl9b, 2s,3s-butane-2,3-diylbis diphenylphosphine, phosphine, 1s,2s-1,2-dimethyl-1,2-ethanediyl bis diphenyl, --chiraphos, 2s,3s-chiraphos
PubChem CID 10113249
IUPAC Name [(2S,3S)-3-diphenylphosphanylbutan-2-yl]-diphenylphosphane
SMILES C[C@@H]([C@H](C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 107°C to 109°C
Quantity 250 mg
Beilstein 4268869
Sensitivity Air Sensitive
Formula Weight 426.48
Percent Purity 98%
Chemical Name or Material (2S,3S)-(-)-Bis(diphenylphosphino)butane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.