missing translation for 'onlineSavingsMsg'
Learn More

(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane, 97%, Thermo Scientific™

Product Code. 11905941 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11905941 100 mg Glass bottle 100mg
11915941 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11905941 Supplier Thermo Scientific Acros Supplier No. 434121000
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1314246-02-0
Molecular Formula C17H25O2P
Molecular Weight (g/mol) 292.36
MDL Number MFCD09265099
InChI Key UKBFKBMQNUOKTD-UHFFFAOYNA-N
Synonym 2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, s,s-et-rajphos, 2-2-2s,5s-2,5-diethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-1,3-dioxolane, trans-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s-1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, 2s,5s---1-2-1,3-dioxolan-2-yl phenyl-2,5-diethylphospholane, kanata purity
PubChem CID 16218309
IUPAC Name 1-[2-(1,3-dioxolan-2-yl)phenyl]-2,5-diethylphospholane
SMILES CCC1CCC(CC)P1C1=CC=CC=C1C1OCCO1

Specifications

Color Colorless to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 mg
Formula Weight 292.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.