missing translation for 'onlineSavingsMsg'
Learn More

(2S)-(+)-Glycidyl tosylate, 99+%

Product Code. 10309490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10309490 5 g Glass bottle 5g
10217170 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10309490 Supplier Thermo Scientific Acros Supplier No. 264580050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 70987-78-9 | C10H12O4S | 228.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70987-78-9
Molecular Formula C10H12O4S
Molecular Weight (g/mol) 228.27
MDL Number MFCD00064489
InChI Key NOQXXYIGRPAZJC-VIFPVBQESA-N
Synonym s-glycidyl tosylate, 2s-+-glycidyl tosylate, s-+-glycidyl tosylate, +-glycidyl tosylate, s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, 2s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, ccris 6400, s-glycidyltosylate, 2s-glycidyl tosylate, oxiranemethanol, 4-methylbenzenesulfonate, s
PubChem CID 153296
IUPAC Name [(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 45°C to 48°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition + 17.00 (20.00°C c=2.5,CHCl3)
Specific Rotation + 17
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in chloroform
Formula Weight 228.27
Percent Purity 99+%
Physical Form Fluffy Powder
Chemical Name or Material (2S)-(+)-Glycidyl tosylate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (2S)-(+)-Glycidyl tosylate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H411-Toxic to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.