missing translation for 'onlineSavingsMsg'
Learn More

(2S)-(+)-Glycidyl Tosylate, TRC

Product Code. 30390627
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390627 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390627 Supplier Toronto Research Chemicals Supplier No. TRCG61600125G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-Glycidyl tosylate
CAS 70987-78-9
Molecular Formula C10 H12 O4 S
Quantity 25 g
Synonym 2-Oxiranemethanol, 2-(4-methylbenzenesulfonate), (2S)-, Oxiranemethanol, 4-methylbenzenesulfonate, (2S)- (9CI), Oxiranemethanol, 4-methylbenzenesulfonate, (S)-, (+)-Glycidyl tosylate, (2S)-(+)-Glycidyl tosylate, (2S)-Glycidyl toslyate, (2S)-Oxiranemethanol 4-methylbenzenesulfonate, (S)-(+)-Glycidyl 4-methylbenzenesulfonate, (S)-(+)-Glycidyl tosylate, (S)-(2,3-Epoxypropan-1-yl) 4-methylbenzenesulfonate, (S)-Glycidyl p-toluenesulfonate, (S)-Glycidyl tosylate, (S)-Oxiranylmethyl toluene-4-sulfonate, (S)-Toluene-4-sulfonic acid oxiranylmethyl ester, 2(S)-Glycidyl tosylate, Toluene-4-sulfonic acid (S)-oxiranylmethyl ester
InChI Formula InChI=1S/C10H12O4S/c1-8-2-4-10(5-3-8)15(11,12)14-7-9-6-13-9/h2-5,9H,6-7H2,1H3/t9-/m0/s1
SMILES Cc1ccc(cc1)S(=O)(=O)OC[C@@H]2CO2
IUPAC Name [(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol) 228.26
Formula Weight 228.0456
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.