missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 2XYT-Broth, Capsules: Regular

Product Code. 11377669 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
454 g
1000 g
5000 g
Format:
Capsule
Unit Size:
1kg
454g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Unit Size
11377669 1000 g Capsule 1kg
11343319 454 g Capsule 454g
11307679 5000 g Capsule 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377669 Supplier MP Biomedicals™ Supplier No. 113012031
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Optimized for the growth and maintenance of M13 phage or other filamentous ssDNA bacteriophages

Regular size capsules.

  • Contents/L: 16g tryptone, 10g yeast extract, 5g NaCl
  • Use 31 capsules/L

Specifications

Certifications/Compliance Molecular Biology Certified™
Content And Storage 15°C to 30°C
Format Capsule
Product Type Microbiology Media
Formulation Tryptone 16g/L, Yeast extract 10g/L, NaCl 5g/L
For Use With (Application) For growth and maintenance of M13 phage or other filamentous phage
Quantity 1000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.