missing translation for 'onlineSavingsMsg'
Learn More

Melford 2XYT Broth, Granulated

Product Code. 30018930
Change view
Click to view available options
Quantity:
500 g
1 kg
2 kg
5 kg
Unit Size:
1kg
2kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018930 1 kg 1kg
30018932 500 g 500g
30018933 5 kg 5kg
30018931 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30018930 Supplier Melford Supplier No. X156801000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nutritionally enriched medium for the growth of recombinant strains of E.

Nutritionally enriched medium for the growth of recombinant strains of E. Coli and in the preparation and transformation of competent cells.

Material is specially granulated to reduce dust and increase solubility rate.

Dissolve 31g in one liter distilled water. Heat solution to boiling with continuous stirring. Sterilize by autoclaving at 121°C for 15 minutes. Final pH 25°C: 6.8-7.2.

Ingredients:

Digest Peptone: 16 g/l

Yeast Extract: 10 g/l

Sodium Chloride: 5 g/l

Specifications

Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.